Ecosense EC30A Handheld Conductivity/TDS/Temperature Pen Tester, YSI
Supplier: YSI
606121
10032-024EA 265.75 USD
10032-024
Ecosense EC30A Handheld Conductivity/TDS/Temperature Pen Tester, YSI
Electrochemistry Meters Conductivity Meters
The EC30A is part of the EcoSense® family of products. The EC30A offers accurate, low-cost measurement of conductivity and TDS (total dissolved solids) along with temperature.
  • Waterproof IP-67
  • Automatic temperature compensation
  • Replaceable electrodes
  • GLP functionality
  • 1- or 2-point calibration
  • Clear, easy-to-read display with on-screen instructions
  • Selectable or auto-ranging for conductivity or TDS
  • Hold feature locks readings on display
  • >200 hour battery life; low battery indicator
Selectable units of measure for conductivity, TDS and temperature.
More Product Information