ClipTip® Pipette Tips, 1250, Thermo Scientific
Supplier: Thermo Fisher Scientific
94410810 94410813 94410817 94410818 94420813 94420818
10030-784CS 423.64 USD
10030-784 10030-786 10030-788 10030-790 10030-792 10030-794
ClipTip® Pipette Tips, 1250, Thermo Scientific
Pipette Tips
Thermo Scientific ClipTip® pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force
  • Feature a hydrophobic inner surface which effectively prevent liquid retention
  • Certified free of RNase, DNase, DNA, ATP, and pyrogen contamination
  • Tips are available in multiple packaging options, including environmentally friendly reloads
ClipTip® 1250 pipette tips are suitable for pipetting 15 to 1250 µl volumes with the E1-ClipTip® electronicMore Product Information