L(+)-Cysteine hydrochloride monohydrate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (R)-(+)-Cysteine hydrochloride monohydrate, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, L-Cysteine HCI Monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, Monohydrate, Cysteine Hydrochloride monohydrate

041081250GM
100278-704EA 129.46 USD
100278-704
L(+)-Cysteine hydrochloride monohydrate ≥99%
L(+)-Cysteine hydrochloride monohydrate
Formula: HSCH₂CH(NH₂)COOH·HCl·H₂O
MDL Number: MFCD00065606
CAS Number: 7048-04-6
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e9d2dd09f1314aea5a750796e1eb24dca4f3c9e&vid=2e9d2dd09f1314aea5a750796e1eb24dca4f3c9e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR