VWR® Advanced 3500 Orbital Shakers, 230 V
Supplier: VWR International
89032-098 10027-144
89032-098EA 4077.25 USD
89032-098 10027-144
VWR® Advanced 3500 Orbital Shakers, 230 V
Shakers and Mixers Orbital Shakers
The VWR® Advanced 3500 Orbital Shaker is designed for a wide range of applications including cell cultures that require accurate and repeatable results. The microprocessor control provides consistent uniform shaking while safely ramping the set speed.
  • 230 V models
  • Exclusive Accu-Drive® shaking system
  • Triple eccentric drive provides reliable service and continuous duty operation
  • Built-in load sensor detects unbalanced conditions and automatically reduces rpm to a safe speed to protect samples
  • Five-year warranty on parts and labor
  • Available with a NIST
More Product Information