VWR® Signature™ Rocking Platform Shaker
Supplier: VWR International
12620-906 10027-104
12620-906EA 1730.78 USD
12620-906 10027-104
VWR® Signature™ Rocking Platform Shaker
Shakers and Mixers Rotating/Rocking Shakers
Ideal for cell culture work, staining and destaining gels, hybridization procedures, hematology, and blotting techniques.
  • Digital display for speed, tilt angle, and time
  • Microprocessor control
  • Timer with audible alarm
  • Electronic tilt tdjustment
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Shakers are designed for use in incubators, CO₂ incubators, cold rooms (–10 to +60 °C), 80% relative humidity,non-condensing environments.
Audible and visualMore Product Information