Zinc sulfide, crystal disk for spectrophotometry, polished

Supplier: INTERNATIONAL CRYSTAL LABS MS

Synonyms: Zinc (II) sulphide, Zinc (II) sulfide

0002I-1858 0002I-4591 0002I-151 0002I-152 0002I-1011 0002I-77 0002I-464
100211-352EA 428.23 USD
100211-352 100214-462 100212-948 100215-600 100211-670 100216-086 100216-122
Zinc sulfide, crystal disk for spectrophotometry, polished
Zinc sulphide

Zinc Sulfide (ZnS) is used as a transmission window for IR spectroscopy. Zinc Sulfide (ZnS) is or has been produced under the trademarks Cleartran® and Irtran® .

Formula: SZn
MW: 97.46 g/mol
Melting Pt: 1064
Density: 4.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011301
CAS Number: 1314-98-3
EINECS: 215-251-3
Merck Index: 12,10294

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR