Trimethyl phosphate ≥97%

Supplier: STREM CHEMICALS INC MS

Synonyms: Trimethylphosphate

93-1561-250G 93-1561-50G
100211-220EA 103.32 USD
100211-220 100197-718
Trimethyl phosphate ≥97%
Trimethyl phosphate

Volatile Precursors for CVD

Formula: C₃H₉O₄P
MW: 140.08 g/mol
Boiling Pt: 197.2
Melting Pt: -46
Density: 1.197
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00008348
CAS Number: 512-56-1
EINECS: 208-144-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR