Iron ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS
Danger

93-2601-1000G 93-2601-250G
100210-684EA 284.49 USD
100210-684 100211-234
Iron ≥99.9%, powder
Iron
Formula: Fe
Boiling Pt: 3000 °C
Melting Pt: 1535 °C
Density: 7,86
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR