Carbon ≥99.999%, powder

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Charcoal

93-0602-5G 93-0602-25G
100210-134EA 35.38 USD
100210-134 100204-130
Carbon ≥99.999%, powder
Carbon
Formula: C
Boiling Pt: 4200 °C
Melting Pt: 3652…3697 °C (subl.)
Density: 1.8…2.1 (amorphous)
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR