1,4,7-Trimethyl-1,4,7-triazacyclononane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1,4,7-Trimethyl-1,4,7-triazonane

07-2750-250MG 07-2750-500MG 07-2750-2G
100200-834EA 191.08 USD
100200-834 100209-560 100198-508
1,4,7-Trimethyl-1,4,7-triazacyclononane ≥97%
1,4,7-Trimethyl-1,4,7-triazacyclononane
Formula: C₉H₂₁N₃
Density: 0.884
Flash Pt: 155 °F
MDL Number: MFCD00012359
CAS Number: 96556-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR