Chromium ≥99.996%, crystallites

Supplier: STREM CHEMICALS INC MS

24-0025-10G 24-0025-50G
100208-860EA 178.34 USD
100208-860 100204-538
Chromium ≥99.996%, crystallites
Chromium
Formula: Cr
Boiling Pt: 2200 °C
Melting Pt: 1890 °C
Density: 7.20
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR