Palladium(II) acetate ≥98% (99.9+%-Pd)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Palladium diacetate, Palladium (II) acetate

46-1780-1G 46-1780-5G
100205-152EA 145.78 USD
100205-152 100199-946
Palladium(II) acetate ≥98% (99.9+%-Pd)
Palladium (II) acetate
Formula: C₄H₆O₄Pd
Melting Pt: 205 °C
MDL Number: MFCD00012453
CAS Number: 3375-31-3
EINECS: 222-164-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR