Bis(ethylbenzene)molybdenum [mixture of (C2H5)xC6H6-x where x = 0-4)]

Supplier: STREM CHEMICALS INC MS
Danger

42-0200-5G 42-0200-1G 42-0200-25G
100203-134EA 355.26 USD
100203-134 100205-120 100199-238
Bis(ethylbenzene)molybdenum [mixture of (C2H5)xC6H6-x where x = 0-4)]
Bis(ethylbenzene)molybdenum

Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Boiling Pt: 150…170 °C (1 mmHg)
MDL Number: MFCD02091699
CAS Number: 32877-00-2
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR