Silver ≥99.99%, shot

Supplier: STREM CHEMICALS INC MS

47-0085-50G 47-0085-10G
100198-850EA 379.32 USD
100198-850 100198-848
Silver ≥99.99%, shot
Silver
Formula: Ag
Boiling Pt: 2212 °C
Melting Pt: 960.5 °C
Density: 10.49
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR