Thulium ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS
Warning

93-6915-5G 93-6915-1G
100198-446EA 884.62 USD
100198-446 100198-444
Thulium ≥99.9%, powder
Thulium
Formula: Tm
Boiling Pt: 1727 °C
Melting Pt: 1545 °C
Density: 9.332
MDL Number: MFCD00011281
CAS Number: 7440-30-4
EINECS: 231-140-2
UN: 3089
ADR: 4,III
Merck Index: 12,09535

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR