Palladium ≥99.95%, powder - 60 mesh

Supplier: STREM CHEMICALS INC MS
Danger

93-4631-1G 93-4631-5G
100196-672EA 270.34 USD
100196-672 100196-674
Palladium ≥99.95%, powder - 60 mesh
Palladium
Formula: Pd
MW: 106.420 g/mol
Boiling Pt: 3140 °C
Melting Pt: 1552 °C
Density: 12.02
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 3089
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR