Gemini Cautery System, Roboz Surgical
Supplier: Roboz Surgical
RS-300
100190-486EA 108 USD
100190-486
Gemini Cautery System, Roboz Surgical
Pens Surgical Markers
The gemini cautery System is the perfect tool for your cauterizing needs. The two tip sizes combined with two different operating temperatures allow for maximum versatility.
  • The tips are very easy to replace
  • Unit uses standard AA batteries
  • Handle has a screw-cap for convenient battery replacement
Using the battery spacer instead of one of the batteries changes the operating temperature from high to low.
Delivery information: Kit contains a handle, two 5 mm tips, two 10 mm tips, two batteries and a battery spacer. The kit is shipped in hard plasticMore Product Information