Dispensette® S Bottle Top Dispensers
Supplier: Brandtech
4600311 4600361 4600310 4600320 4600330 4600340 4600350 4600360 4600101 4600121 4600131 4600141 4600151 4600161 4600171 4600100 4600160 4600170 4600211 4600221 4600231 4600241 4600210 4600220 4600230 4600240 4600120 4600130 4600140 4600150 4600321 4600331 4600341 4600351
10018-930EA 927.46 USD
10018-930 10018-932 10018-934 10018-936 10018-938 10018-940 10018-942 10018-944 10018-946 10018-948 10018-950 10018-952 10018-954 10018-956 10018-958 10018-960 10018-962 10018-964 10018-966 10018-968 10018-970 10018-972 10018-974 10018-976 10018-978 10018-980 10019-010 10019-012 10019-014 10019-016 10020-960 10020-962 10020-964 10020-966
Dispensette® S Bottle Top Dispensers
Bottle Top Dispensers
Fixed volume or variable volume, analogue-adjustable or digital easy calibration.
  • With or without recirculation valve
  • Autoclavable at 121 °C
  • Three different models of volume adjustment available: Fixed (fixed volume for standard applications; simple calibration adjustment with supplied tool) and Analogue (analogue volume display, fast volume adjustment; simple calibration adjustment with supplied tool)
  • Digital EASY CALIBRATION: Easy to read digital display; dispensing volume can be set accurately and reproducibly. Easy calibration technique for adjustment
More Product Information