Permanent Polyester Labels, Type B30, ToughBond®, Brady
Supplier: Brady Worldwide
145295
10018-036EA 328.31 USD
10018-036
Permanent Polyester Labels, Type B30, ToughBond®, Brady
Labels
Permanent Polyester (B-484) material has excellent pliability and is designed for high adhesion to textured metals, low surface energy plastics, and angled or curved surfaces.
  • Ideal for marking backer board, DIN rail, relays, PLCs, switches, solenoids, drive motors, and other components inside panels
  • Glossy white polyester with permanent rubber based adhesive is thermal transfer printable
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved
  • Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll