Permanent Polyester Labels, Type B30, WorkHorse®, Brady
Supplier: Brady Worldwide
143578
10017-986EA 285.15 USD
10017-986
Permanent Polyester Labels, Type B30, WorkHorse®, Brady
Labels
Permanent polyester label (B-423) is ideal for electronic PCB and component identification, barcode labels, rating plates, and solar panel identification.
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted.
Meets requirements of a halogen-free material per DIN VDE 0472 part 815.
Order Now
SPECIFICATIONS
Environmentally Preferable