SlideMate™ AS Slide Printer, Epredia™
Supplier: Epredia
A83900001 A83900004 A83900002 A83900005
10754-528EA 15207.73 USD
10754-528 10754-530 10017-188 10017-190
SlideMate™ AS Slide Printer, Epredia™
Printers Cassette and Slide Printers
Reduce specimen identification errors while enhancing lean workflow with the Epredia™ SlideMate™ AS On-Demand Slide Printer.
  • Fully integrated color touch-screen and simple software are easy to use
  • Build-in barcode scanner reads cassette information and imports the data to the slide label
  • Connects easily to other computers or Laboratory Information Systems
  • Prints directly onto the slide, eliminating the need for hand writing or labels
  • 1 roll capacity, approximately 12,000 slides per roll
The SlideMate™ AS is the next generation Epredia™ Slide PrinterMore Product Information