Pipetting Reservoirs, Argos Technologies
Supplier: Antyila Scientific
04395-27 04395-28 04395-29 04395-30 04395-23 04395-24 04395-25 04395-31
10015-224CS 276.93 USD
10015-224 10015-226 10015-228 10015-230 10015-232 10015-234 10015-236 10015-240
Pipetting Reservoirs, Argos Technologies
Reagent Reservoirs
These reagent reservoirs are created to work with 8- and 12-channel pipettes
The thick-wall construction of each basin provides a noticeable difference in product and performance. Low cross bars on each end of the basin give the product much more rigidity and durability than competitive offerings.
The reservoirs were developed for ease of use. They feature pour spouts in every corner, and one larger spout on each reservoir to facilitate spill-free pouring. In addition, the individually wrapped and 5-pack versions are packaged in easy-open “Snap & Tear” bags. No need for scissors,More Product Information