Reagent alcohol ≥95%, Surgipath® ISO for histology and pathology

Supplier: Leica Biosystems
Danger

Synonyms: Reagent alcohol

3803682
10015-097CS 197.57 USD
10015-097
Reagent alcohol ≥95%, Surgipath® ISO for histology and pathology
Alcohol

Ethanols are pure filtered and laboratory tested to ensure a consistent high quality for all formulating, processing and staining procedures in histology and cytology.


  • High-quality filtered Ethanol blend
  • ISO certified
  • Available in non‑breakable, gallon containers


Certifications: ISO certified

Flash Pt: 55 °F
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR