Hematoxylin solution_SC Gill III staining_nuclear Hg_free

Supplier: Leica Biosystems
Warning

Synonyms: DcH-48, Natural Black I, Hydroxybrasilin

3801540
10015-070GL 182.69 USD
10015-070
Hematoxylin solution_SC Gill III staining_nuclear Hg_free
Hematoxylin staining solutions

The Gill formula hematoxylins are used in the progressive method of staining.


Three Gill formulations are available:
The Gill I formula is generally recommended for cytology.
The stronger Gill III is usually preferred for use in histology.
GIll II is a medium strength hematoxylin.

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR