Surgipath® Sectioning Blades, Leica
Supplier: Leica Biosystems
14035843489 14035843488 14035843497 14035838382 14035838925 14035838926 14035843490 14035843491 14035838383
10158-934CS 2966.96 USD
10158-934 10158-936 10158-938 10015-010 10015-014 10015-016 10015-022 10015-024 10015-030 10015-142 15147-841
Surgipath® Sectioning Blades, Leica
Microtomes and Microtome Assemblies Microtome Blades and Handles
Disposable low-profile and high-profile microtome blades for routine sectioning.
  • Stainless steel construction
  • Ultra-fine blade
  • Durable
  • Coating provides higher-quality sectioning ribbons
Low-Profile 819 Disposable Blades are 80mm in length, 8mm high, and 0.25mm thick
Made of stainless steel, these blades are ultra-fine yet durable. They can be used with the Leica microtome and cryostat series, as well as for common cryostat and microtome models. An inventive blade coating process provides perceptibly improved results in section stretching and in theMore Product Information