Scalpel Blades, Leica
Supplier: Leica Biosystems
3802160 3802181 3802186 3802187 3802182
10014-988BX 119.33 USD
10014-988 10014-990 10014-992 10014-994 10014-996
Scalpel Blades, Leica
Scalpels and Scalpel Assemblies Scalpel Blades
The Feather Autopsy, Trimming, and Dissection range is internationally renowned for its sharpness and versatility.
  • Stainless steel or carbon steel construction
  • Scalpel tips are available in a variety of formats
  • Supplied sterile or nonsterile
  • Excellent feel and dexterity
The #22 feather brand scalpel blades are stainless steel, which provide a durable, precision edge
A diverse range of scalpel blades and handles for use in the pathology laboratory and mortuary are also available. The #22 and #60 scalpels are available in the traditional pointed tipMore Product Information