Axygen® Microcentrifuge Screw Caps, Corning
Supplier: Corning
SCO-B SCO-BR SCO-C SCO-G SCO-LP-B SCO-LP-C SCO-LP-Y SCO-R SCO-V SCO-Y
10011-428CS 961.39 USD
10011-428 10011-430 10011-432 10011-434 10011-440 10011-442 10011-444 10011-460 10011-466 10011-468 10011-472
Axygen® Microcentrifuge Screw Caps, Corning
Closures Test/Sample Tube Closures
These caps are compatible with all Axygen Scientific screw top microcentrifuge tubes, including both skirted and nonskirted variations of the 0.5 ml, 1.5 ml, and 2 ml microcentrifuge tubes.
  • Available looped and non-looped
The O-ring closure is formulated and manufactured from a special compound resistant to chemicals such as anhydrous ethyl alcohol, purified ether, hexane, acetone, chloroform, acetonitrile, 40% trifluoroacetic acid solution, and other chemicals used in most laboratories. The caps have extra long lips to help protect samples and ribsMore Product Information