Bottle Holders, White ABS, TrippNT
Supplier: TrippNT
50080 50016 50017 50011 50411 50085 50157 50189
10010-964EA 43.65 USD
10010-964 11376-326 11376-328 11376-330 10010-968 10010-966 10010-970 10010-972
Bottle Holders, White ABS, TrippNT
Carriers Bottle Carriers/Containers
Store, secure and stabilize top heavy bottles with resistant bottle holders in 4 sizes. Store 500 or 1000 ml bottles securely and cleanly.
  • White chemically resistant ABS construction
  • Keeps bottles upright and secure
  • Stabilize top heavy plastic bottles
  • In stock and ready to ship
  • American made by TrippNT
  • Total replacement lifetime guarantee
Always know where to put them and when they need to be filled or replaced. This exceptionally handy holder keeps bottles secure and upright. Bright white ABS construction is chemically resistant and provides a clean,More Product Information