VWR® Snap-Cap Centrifuge Tube, 5 ml
Supplier: VWR International
10002-728 10002-740 10002-731
10002-728PK 84.65 USD
10002-728 10002-740 10002-731
VWR® Snap-Cap Centrifuge Tube, 5 ml
Tubes Centrifuge Tubes
VWR® Snap-Cap Centrifuge tubes are made of high-clarity, low-binding, virgin polypropylene.
  • Constructed of high clarity, low-binding, virgin polypropylene
  • Tubes can withstand up to 25000 ×g
  • Supplied with an attached cap with a tight, leak-proof seal
  • Certified RNase- and DNase-free
The tubes have a 16 mm Ø, which is identical to standard 15 ml tubes. This ensures compatibility with existing centrifuge rotors.
Tubes feature dual gradations at every 0.2 and 0.5 ml level. They are supplied with an attached cap that provides a tight, leak-proof seal.More Product Information