Analytical Chemistry Solutions
Avantor Analytical Chemistry
No matter what methods or technology you use, Avantor offers a comprehensive catalog of the equipment, reagents, chemicals, consumables, tools, and other products you need every day. We have everything labs need for industrial, academic or government applications.
Analytical Chemistry solutions
Avantor carries a broad range of products to meet the specialized needs of analytical chemistry laboratories. We offer high-quality products from the world’s most trusted brands to help you and your colleagues answer the most challenging questions in biomedical research, environmental and forensic work, clinical diagnostics, and other materials science applications.
Mass Spectrometry
Mass spectrometry (MS) is an important tool when specific compounds and materials must be identified with a high degree of precision. It is widely utilized in food quality and safety, drug testing, analyzing pesticide residue, and protein identification.
High Performance Liquid Chromatography
High-performance liquid chromatography (HPLC) and ultra-high-performance liquid chromatography (UHPLC) are among the most widely used analytical techniques in the chemical and life sciences.
Low Pressure Chromatography
Preparative or purification chromatography is used to isolate compounds in quantities and at purity levels needed for further experiments or processes by separating them from starting materials, side reactions or other impurities.
Gas Chromatography
Gas chromatography (GC) employs insert gases to separate, identify, and quantify volatile sample mixtures in food, environmental, and chemical industries. Avantor carries a variety of columns, solvents, consumables and gas generators
Atomic Spectroscopy
Atomic spectroscopy encompasses a group of analytical techniques based on how atoms absorb and emit electromagnetic radiation, including atomic absorption spectroscopy (AA) and inductively coupled plasma (ICP)
Molecular Spectroscopy
Molecular spectroscopy represents a collection of analytical techniques based on how molecules absorb light, including ultraviolet/visible (UV/Vis), infra-red (IR), Fluorescence, and NMR that are routinely used in key applications within pharmaceuticals
Titration
Titration refers to quantitative chemical analysis based on a reaction between a component of a sample and another substance with which that component of interest reacts in a definite, known proportion.
Karl Fischer water determination
Karl Fischer titration (KF) is the leading method for measuring water content in a wide variety of products and industries, such as chemicals, petroleum products, foods, pharmaceuticals, and many others.
pH & Conductivity Measurement
These electrochemical techniques provide powerful and versatile analytical options that feature high sensitivity, accuracy, and precision, utilizing relatively lower cost instrumentation than other methods of analysis.
What Is Analytical Chemistry? 
Analytical chemistry is a scientific process by which scientists determine the chemical structure and amounts of compounds. Scientists use classic analytical chemistry techniques and advanced equipment to separate, identify, and quantify matter. 
Many analytical chemistry applications span various industries, including pharmaceuticals, food, agriculture, and more.  
Some of the most common analytical chemistry applications include: 
  • Clinical analysis 
  • Forensic analysis 
  • Biomedical research 
  • Pharmaceutical analysis 
  • Food and beverage analysis 
  • Agricultural product quality and stability 
  • Environmental monitoring 
  • Industrial manufacturing quality control