Anti-GAPDH Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A22787-100UG A22787-50UG
77737-953EA 330 USD
77737-953 77855-801
Anti-GAPDH Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: GAPDH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cricetulus griseus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR