Anti-pbpA Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A51926-100UG A51926-50UG
77818-086EA 330 USD
77818-086 77818-087
Anti-pbpA Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: pbpA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Clostridium botulinum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR