2-Ethoxybenzamide ≥98.0%

Supplier: TCI America

Synonyms: Ethenzamide

E0222-500G E0222-25G
TCE0222-025GEA 23.82 USD
TCE0222-025G TCE0222-500G
2-Ethoxybenzamide ≥98.0%
2-Ethoxybenzamide

CAS Number: 938-73-8
MDL Number: MFCD00007977
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 131


Other Product Names / Synonyms: 2-Ethoxybenzoylamide

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 131 °C
MDL Number: MFCD00007977
CAS Number: 938-73-8
EINECS: 213-346-4
https://us.vwr.com/stibo/web/std.lang.all/77/82/28967782.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR