2,4-Diaminophenol dihydrochloride ≥97.0% (by total nitrogen and titration analysis)
Supplier: TCI America
D0080-500G
D0080-25G
TCD0080-500GEA
380.55
USD
TCD0080-500G
TCD0080-25G
2,4-Diaminophenol dihydrochloride ≥97.0% (by total nitrogen and titration analysis)
2,4-Diaminophenol dihydrochloride
CAS Number: 137-09-7
MDL Number: MFCD00012979
Molecular Formula: C6H8N2O
Molecular Weight: 197.06
Purity/Analysis Method:
Product Categories: Bioscience, Cosmetic Ingredients for Research and Experimental Use
Other Product Names / Synonyms: 2,4-Diaminophenol Dihydrochloride
Formula:
C₆H₁₀Cl₂N₂O Melting Pt: 205 °C |
MDL Number:
MFCD00012979 CAS Number: 137-09-7 EINECS: 205-279-4 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...