Sporicidin® disinfectant wipes and towelettes are EPA registered disinfectant wipes recommended for use in hospitals, and medical, dental, and veterinary clinics. Sporicidin® Disinfectant towelettes are used for hospital disinfecting, compliant for medical instrument disinfecting prior to sterilization.
- Broad spectrum disinfectant, with no rotation of intermediate disinfectants needed
- EPA toxicity rating "low/very low"
- Cleans, disinfects, and deodorizes in one step
- Kills bacteriostatic activity up to 6 months, if not removed from the surface
- Mildly alkaline (pH 8.5); fungicidal, bactericidal, virucidal, mildewcidal, and tuburculocidal
- Non-staining, non-flammable, and non-abrasive to most surfaces
- Non-hazardous formula with no special shipping or disposal restrictions
Non-corrosive to surfaces including plastics, latex, vinyl, glass, treated wood, metal, and glazed porcelain. Wipes contain a pharmaceutical active ingredient system with no harsh chemicals, such as bleach and alcohol and are safe for medical and dental equipment.
Disposable wipes are available in multiple sizes for a variety of applications. The standard canister size, 12.7×20.3 cm (5×8"), is ideal for office or lab use. These portable and handy wipes are packaged in 180-count canisters.
The jumbo canister size, 30.5×24 cm (12×9.5"), is designed for heavy-duty cleaning needs, and have two distinct cleaning surfaces. One side has an abrasive texture for aggressive cleaning, while the other side provides softer texture for gentle, delicate cleaning.
Certifications: Meet OSHA’s Bloodborne Pathogens Standards (29 CFR 1910.1030). EPA registered in all 50 states (including California) and Puerto Rico.