Aluminum etch

Supplier: Thermo Scientific Chemicals
Danger

044581-LV 044581-R0
AA44581-R0EA 98.85 USD
AA44581-R0 AA44581-LV
Aluminum etch
Aluminum etch

Applications: Standard aluminum etchant for use on silicon devices and other microelectronic applications

Storage Temperature: Ambient
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR