Stigmasterol ≥90.0%
Supplier: TCI America
S0088-1G
S0088-5G
S0088-25G
TCS0088-001GEA
52.39
USD
TCS0088-001G
TCS0088-025G
TCS0088-5G
Stigmasterol ≥90.0%
Stigmasterol
CAS Number: 83-48-7
MDL Number: MFCD00003630
Molecular Formula: C29H48O
Molecular Weight: 412.70
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 167
Specific rotation [a]20/D: -50.5 deg (C=2, CHCl3)
Lambda max.: 226 nm (MeOH)
Formula:
C₂₉H₄₈O Melting Pt: 167 °C |
MDL Number:
MFCD00003630 CAS Number: 83-48-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...