N-Glycolylneuraminic acid ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Neu5Gc

G0336-10MG
TCG0336-10MGEA 464.51 USD
TCG0336-10MG
N-Glycolylneuraminic acid ≥98.0% (by HPLC)
N-Glycolylneuraminic acid

CAS Number: 1113-83-3
MDL Number: MFCD00057551
Molecular Formula: C11H19NO10
Molecular Weight: 325.27
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Storage Temperature: 0°C


Product Categories: Bioscience, Sialic Acids
Other Product Names / Synonyms: Neu5Gc

Formula: C₁₁H₁₉NO₁₀
MW: 325.27 g/mol
MDL Number: MFCD00057551
CAS Number: 1113-83-3
https://us.vwr.com/stibo/web/std.lang.all/99/45/28969945.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR