10-Undecenal ≥94.0%

Supplier: TCI America

Synonyms: 10-Undecenyl aldehydeUndec-10-enalUndecylenic aldehyde10-Undecenal

U0011-25ML
TCU0011-025MLEA 46.57 USD
TCU0011-025ML
10-Undecenal ≥94.0%
10-Undecenal

CAS Number: 112-45-8
MDL Number: MFCD00007032
Molecular Formula: C11H20O
Molecular Weight: 168.28
Purity/Analysis Method: 94.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 103
Flash Point (°C): 100
Specific Gravity (20/20): 0.84


Other Product Names / Synonyms: 10-Undecenyl Aldehyde, Undecylenic Aldehyde

Formula: C₁₁H₂₀O
MW: 168.28 g/mol
Boiling Pt: 103 °C
Melting Pt: 7 °C
Density: 0.84 (20 °C)
Flash Pt: 100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007032
CAS Number: 112-45-8
EINECS: 203-973-1
https://us.vwr.com/stibo/web/std.lang.all/71/77/28997177.jpg

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR