Mercury(II) chloride USP Test Solution (TS)

Supplier: Ricca Chemical

Synonyms: Mercury dichlorideMercuric chlorideMercury Chloride

R4660000-500A
RC4660-16EA 188.99 USD
RC4660-16
Mercury(II) chloride USP Test Solution (TS)
Mercury (II) chloride
Formula: HgCl₂
MW: 271.5 g/mol
MDL Number: MFCD00011041
CAS Number: 7487-94-7
UN: 1624
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR