2-Chloro-4-iodopyridine ≥95%

Supplier: Matrix Scientific

009790-25G 009790-5G
101797-270EA 138.46 USD
101797-270 101797-268
2-Chloro-4-iodopyridine ≥95%
2-Chloro-4-iodopyridine

Hazard code(s): Irritant, Store in Freezer

Formula: C₅H₃ClIN
MW: 239.44 g/mol
Melting Pt: 41.2…41.3 °C
MDL Number: MFCD01861983
CAS Number: 153034-86-7

Order Now

Specification Test Results

Melting point 41.2-41.3°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR