Ethyl-4-iodobenzoate ≥98%
Supplier: Matrix Scientific
Synonyms:
Ethyl 4-iodobenzoate
, 4-Iodobenzoic acid ethyl ester
004868-25G
004868-100G
101787-930EA
73.85
USD
101787-930
101787-928
Ethyl-4-iodobenzoate ≥98%
Ethyl 4-iodobenzoate
Hazard code(s): Irritant, Light Sensitive
Formula:
C₉H₉IO₂ MW: 276.07 g/mol Boiling Pt: 112 °C (1mm) Density: 1.630 g/cm³ |
MDL Number:
MFCD00017344 CAS Number: 51934-41-9 |
Specification Test Results
Boiling point | 112°C/1mm |
Density | 1.63 |
ND20 | 1.588 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...