4-Formylphenylboronic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: (4-Formylphenyl)metaboric acid4-Formylbenzeneboronic acid4-BoronobenzaldehydeDihydroxy(4-formylphenyl)borane

003917-25G
101786-452EA 46.15 USD
101786-452
4-Formylphenylboronic acid ≥97%
4-Formylphenylboronic acid

Hazard code(s): Irritant, Air Sensitive, Store in Freezer

Formula: C₇H₇BO₃
MW: 149.94 g/mol
Melting Pt: 265…268 °C
MDL Number: MFCD00151823
CAS Number: 87199-17-5

Order Now

Specification Test Results

Melting point 265-268°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR