Ward's® Symbiosis Lab Activity
Supplier: VWR
470007-318KT 118.29 USD
470007-318
Ward's® Symbiosis Lab Activity
Specimens Live Specimens Protists - Living Material
Explore Different Examples of Symbiosis
  • Demonstrates parasitism and mutualism
  • Observe fungi, microorganisms, plants, and insects
  • Includes a teacher's guide
In one lab activity students can observe mutualism between fungus and algae, between plant and bacteria, and among protists. They can also observe parasitism between a protozoan and beetle larva. It comes with materials for 30 students and a teacher’s guide.
Note: Coupon included for perishable materials. Redeem by phone or e-mail.