Potassium bromide, powder for spectrophotometry

Supplier: INTERNATIONAL CRYSTAL LABS MS
Warning

0011-5078
100216-064EA 229.46 USD
100216-064
Potassium bromide, powder for spectrophotometry
Potassium bromide

KBr powder is graded solely by purity standards that are relevant to infrared spectroscopy. We do not use such terms as 5 9s (99.999%) , 6 9s (99.9999%) as the means used to test material for such certifications, such as emission spectroscopy, do not dete

Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435 °C (1013 hPa)
Melting Pt: 730 °C
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR