Potassium chloride, powder for spectrophotometry

Supplier: INTERNATIONAL CRYSTAL LABS MS

Synonyms: Potassiumchloride

0011-4129
100215-936EA 148.94 USD
100215-936
Potassium chloride, powder for spectrophotometry
Potassium chloride

KBr powder is graded solely by purity standards that are relevant to infrared spectroscopy. We do not use such terms as 5 9s (99.999%) , 6 9s (99.9999%) as the means used to test material for such certifications, such as emission spectroscopy, do not dete

Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR