HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥97%

Supplier: 0000018203
Danger

Synonyms: Hexamethyldisilazane

93-1413-100G 93-1413-500G
100198-306EA 198.15 USD
100198-306 100196-504
HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥97%
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)
Formula: (CH₃)₃SiNHSi(CH₃)₃
MW: 161.39 g/mol
Boiling Pt: 126 °C
Melting Pt: –82 °C
Density: 0.774
Flash Pt: 48 °F
Storage Temperature: Ambient
MDL Number: MFCD00008259
CAS Number: 999-97-3
EINECS: 213-668-5
UN: 1993
ADR: 3,II
REACH: 01-2119438176-38
Merck Index: 13,04708
https://us.vwr.com/stibo/web/std.lang.all/80/76/2298076.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR