Zinc selenide, crystal optic disc 13 mm x 1 mm, polished both sides
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Zinc (II) selenide
41883-KT
041883-KT
AA41883-KTEA
247
USD
AA41883-KT
CAAA41883-KT
Zinc selenide, crystal optic disc 13 mm x 1 mm, polished both sides
Zinc selenide
Common Applications: Spectroscopy, IR window
Formula:
SeZn MW: 144.35 g/mol Melting Pt: 1793 °C Density: 5.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011296 CAS Number: 1315-09-9 EINECS: 215-259-7 UN: 3283 ADR: 6.1,III Merck Index: 12,10290 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...