Calcium fluoride, crystal optic disc 13 mm x 2 mm, polished both sides
Supplier: Thermo Scientific Chemicals
|
Warning
|
38428-KT
038428-KT
AA38428-KTEA
150
USD
AA38428-KT
CAAA38428-KT
Calcium fluoride, crystal optic disc 13 mm x 2 mm, polished both sides
Calcium fluoride
Formula:
CaF₂ MW: 78.07 g/mol Boiling Pt: 2500 °C (1013 hPa) Melting Pt: 1418 °C Density: 3.18 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010907 CAS Number: 7789-75-5 EINECS: 232-188-7 Merck Index: 13,01669 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...