Anti-CTLA4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48956-100UG A48956-50UG
77705-379EA 253.75 USD
77705-379 77715-566
Anti-CTLA4 Rabbit Polyclonal Antibody
Antibodies
IHC:1:50-300, ELISA:1:10000-20000

Type:
Antigen: CTLA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR